siRNA Details
| Gene Symbol: NOC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022756.1 | - | 12 | 1440 | ||
| AK092491.1 | - | 4 | 994 | |||
| AK315080.1 | - | 18 | 2166 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF161376.1 | - | 1 | 70 | Chr. 1 - 925150 - 945132 [-] on Build GRCh38 | |

NOC2L
Genomic Map