siRNA Details
| Gene Symbol: SAMD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BM055062.1 | - | 1 | 375 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF161376.1 | - | 2, 3 | 684 | Chr. 1 - 925150 - 945132 [-] on Build GRCh38 | |

SAMD11
Genomic Map