siRNA Details
| Gene Symbol: SOX9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056229.1 | - | 2 | 1155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX9-AS1| Gene Symbol: SOX9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056229.1 | - | 2 | 1155 | ||