siRNA Details
Gene Symbol: LOC100996455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073177.1 | 1 | 417 | |||
GenBank mRNA | AK094364.1 | - | 1 | 402 | ||
HY146585.1 | - | 1 | 418 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073177.1 | 1 | 417 | |||
GenBank mRNA | AK094364.1 | - | 1 | 402 | ||
HY146585.1 | - | 1 | 418 |