siRNA Details
| Gene Symbol: LOC100996455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073177.1 | 2 | 800 | |||
| GenBank mRNA | AK094364.1 | - | 2 | 785 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996455| Gene Symbol: LOC100996455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073177.1 | 2 | 800 | |||
| GenBank mRNA | AK094364.1 | - | 2 | 785 | ||