siRNA Details
| Gene Symbol: LINC00641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038971.1 | 1 | 193 | |||
| GenBank mRNA | BX537733.1 | - | 1 | 193 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555688.1 | - | 1 | 193 | Chr. 14 - 21200461 - 21206900 [-] on Build GRCh38 | |

LINC00641
Genomic Map