siRNA Details
Gene Symbol: LINC00641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038971.1 | 1 | 236 | |||
GenBank mRNA | BX537733.1 | - | 1 | 236 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000555688.1 | - | 1 | 236 | Chr. 14 - 21200461 - 21206900 [-] on Build GRCh38 |
