siRNA Details
| Gene Symbol: DOCK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF339774.1 | - | 1 | 242 | ||
| AK022011.1 | - | 1 | 1393 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DOCK9| Gene Symbol: DOCK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF339774.1 | - | 1 | 242 | ||
| AK022011.1 | - | 1 | 1393 | |||