siRNA Details
| Gene Symbol: DOCK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF339774.1 | - | 1 | 424 | ||
| AK022011.1 | - | 1 | 1575 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX100976.1 | - | 1 | 424 | Chr. 13 - 98849432 - 98850065 [-] on Build GRCh38 | |

DOCK9
Genomic Map