siRNA Details
| Gene Symbol: LINC00309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033837.1 | 1 | 93 | |||
| GenBank mRNA | AL137337.1 | - | 1 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00309| Gene Symbol: LINC00309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033837.1 | 1 | 93 | |||
| GenBank mRNA | AL137337.1 | - | 1 | 93 | ||