siRNA Details
Gene Symbol: LINC00309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033837.1 | 6 | 2792 | |||
GenBank mRNA | AL137337.1 | - | 6 | 2792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431020.1 | - | 6 | 2792 | Chr. 2 - 64185078 - 64205485 [-] on Build GRCh38 |
