siRNA Details
| Gene Symbol: LOC1720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033423.1 | 1 | 437 | |||
| GenBank mRNA | J00146.1 | - | 1 | 434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC1720| Gene Symbol: LOC1720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033423.1 | 1 | 437 | |||
| GenBank mRNA | J00146.1 | - | 1 | 434 | ||