siRNA Details
Gene Symbol: LOC101927513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134297.1 | 1 | 966 | |||
GenBank mRNA | BC034804.1 | - | 1 | 966 | ||
DB519943.1 | - | 1 | 18 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134297.1 | 1 | 966 | |||
GenBank mRNA | BC034804.1 | - | 1 | 966 | ||
DB519943.1 | - | 1 | 18 |