siRNA Details
| Gene Symbol: COMMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057145.1 | - | 2 | 780 | ||
| AK293708.1 | - | 4 | 338 | |||
| BC033744.1 | - | 2 | 799 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124968.1 | - | 1 | 2751 | Chr. 15 - 75336063 - 75340266 [+] on Build GRCh38 | |

COMMD4
Genomic Map