siRNA Details
| Gene Symbol: LOC101927919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110854.1 | 3 | 497 | |||
| GenBank mRNA | AK125564.1 | - | 3 | 497 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA896292.1 | - | 3 | 497 | Chr. 6 - 117673953 - 117675284 [-] on Build GRCh38 | |

LOC101927919
Genomic Map