siRNA Details
| Gene Symbol: LOC101927919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110854.1 | 3 | 1528 | |||
| GenBank mRNA | AK125564.1 | - | 3 | 1528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927919| Gene Symbol: LOC101927919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110854.1 | 3 | 1528 | |||
| GenBank mRNA | AK125564.1 | - | 3 | 1528 | ||