siRNA Details
| Gene Symbol: ZNF99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080409.2 | NP_001073878.2 | 4 | 7625 | ||
| GenBank mRNA | BC021822.1 | - | 1 | 2437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF99| Gene Symbol: ZNF99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080409.2 | NP_001073878.2 | 4 | 7625 | ||
| GenBank mRNA | BC021822.1 | - | 1 | 2437 | ||