siRNA Details
Gene Symbol: DBIL5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036635.2 | 1 | 1360 | |||
GenBank mRNA | AF229803.1 | - | 1 | 1360 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DBIL5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036635.2 | 1 | 1360 | |||
GenBank mRNA | AF229803.1 | - | 1 | 1360 |