siRNA Details
| Gene Symbol: DBIL5P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036635.2 | 1 | 148 | |||
| GenBank mRNA | AF229803.1 | - | 1 | 148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DBIL5P2| Gene Symbol: DBIL5P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036635.2 | 1 | 148 | |||
| GenBank mRNA | AF229803.1 | - | 1 | 148 | ||