siRNA Details
| Gene Symbol: LOC541472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131935.1 | 2 | 1145 | |||
| GenBank mRNA | BC027978.1 | - | 2 | 1151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC541472| Gene Symbol: LOC541472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131935.1 | 2 | 1145 | |||
| GenBank mRNA | BC027978.1 | - | 2 | 1151 | ||