siRNA Details
Gene Symbol: LOC541472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131935.1 | 1 | 17 | |||
GenBank mRNA | BC027978.1 | - | 1 | 23 | ||
BI912463.1 | - | 1 | 17 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC541472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131935.1 | 1 | 17 | |||
GenBank mRNA | BC027978.1 | - | 1 | 23 | ||
BI912463.1 | - | 1 | 17 |