siRNA Details
| Gene Symbol: LINC00667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119664.1 | - | 1 | 30 | ||
| BC119665.1 | - | 1 | 30 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00667| Gene Symbol: LINC00667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119664.1 | - | 1 | 30 | ||
| BC119665.1 | - | 1 | 30 | |||