siRNA Details
| Gene Symbol: LOC101928902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134307.1 | 2 | 1106 | |||
| GenBank mRNA | BC026857.1 | - | 2 | 590 | ||
| BC047297.1 | - | 2 | 1120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928902| Gene Symbol: LOC101928902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134307.1 | 2 | 1106 | |||
| GenBank mRNA | BC026857.1 | - | 2 | 590 | ||
| BC047297.1 | - | 2 | 1120 | |||