siRNA Details
Gene Symbol: C5orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514089.2 | XP_011512391.1 | 53 | 10321 | ||
GenBank mRNA | AK022380.1 | - | 1 | 639 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU122286.2 | - | 1 | 640 | Chr. 5 - 37079097 - 37079942 [-] on Build GRCh38 |
