siRNA Details
| Gene Symbol: C5orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514089.2 | XP_011512391.1 | 53 | 10003 | ||
| GenBank mRNA | AK022380.1 | - | 1 | 321 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU122286.2 | - | 1 | 321 | Chr. 5 - 37079097 - 37079942 [-] on Build GRCh38 | |

C5orf42
Genomic Map