siRNA Details
| Gene Symbol: GAPLINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110428.1 | 3 | 445 | |||
| NR_110429.1 | 3 | 471 | ||||
| GenBank mRNA | AJ707365.1 | - | 3 | 408 | ||
| BM782281.1 | - | 3 | 445 | |||
| BX107736.1 | - | 3 | 383 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAPLINC| Gene Symbol: GAPLINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110428.1 | 3 | 445 | |||
| NR_110429.1 | 3 | 471 | ||||
| GenBank mRNA | AJ707365.1 | - | 3 | 408 | ||
| BM782281.1 | - | 3 | 445 | |||
| BX107736.1 | - | 3 | 383 | |||