siRNA Details
| Gene Symbol: TMEM43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007176.1 | XP_016862665.1 | 2 | 357 | ||
| GenBank mRNA | AK026869.1 | - | 1 | 1280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM43| Gene Symbol: TMEM43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007176.1 | XP_016862665.1 | 2 | 357 | ||
| GenBank mRNA | AK026869.1 | - | 1 | 1280 | ||