siRNA Details
| Gene Symbol: MIPOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX647795.1 | - | 10 | 5119 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK127356.1 | - | 1 | 3305 | Chr. X - 108629397 - 108633451 [+] on Build GRCh38 | |

MIPOL1
Genomic Map