siRNA Details
| Gene Symbol: CAMKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128601.1 | - | 8 | 3119 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126617.1 | - | 1 | 1116 | Chr. 17 - 3878826 - 3880725 [-] on Build GRCh38 | |

CAMKK1
Genomic Map