siRNA Details
| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122122.1 | 2 | 1029 | |||
| GenBank mRNA | AB050001.1 | - | 1 | 262 | ||
| AB050003.1 | - | 2 | 1028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSL| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122122.1 | 2 | 1029 | |||
| GenBank mRNA | AB050001.1 | - | 1 | 262 | ||
| AB050003.1 | - | 2 | 1028 | |||