siRNA Details
| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122122.1 | 3 | 1194 | |||
| NR_122123.1 | 2 | 974 | ||||
| GenBank mRNA | AB050001.1 | - | 2 | 427 | ||
| AB050002.1 | - | 2 | 302 | |||
| AB050003.1 | - | 3 | 1193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSL| Gene Symbol: TSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122122.1 | 3 | 1194 | |||
| NR_122123.1 | 2 | 974 | ||||
| GenBank mRNA | AB050001.1 | - | 2 | 427 | ||
| AB050002.1 | - | 2 | 302 | |||
| AB050003.1 | - | 3 | 1193 | |||