siRNA Details
| Gene Symbol: ZNF283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026636.1 | XP_016882125.1 | 1 | 1993 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK123399.1 | - | 1 | 464 | Chr. 19 - 43842059 - 43844242 [+] on Build GRCh38 | |

ZNF283
Genomic Map