siRNA Details
| Gene Symbol: LOC100996437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110037.1 | 2 | 309 | |||
| GenBank mRNA | BM826587.1 | - | 2 | 277 | ||
| HY006622.1 | - | 3 | 398 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996437| Gene Symbol: LOC100996437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110037.1 | 2 | 309 | |||
| GenBank mRNA | BM826587.1 | - | 2 | 277 | ||
| HY006622.1 | - | 3 | 398 | |||