siRNA Details
Gene Symbol: LOC101928907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135690.1 | 2 | 1771 | |||
GenBank mRNA | BC034325.1 | - | 1 | 1879 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135690.1 | 2 | 1771 | |||
GenBank mRNA | BC034325.1 | - | 1 | 1879 |