siRNA Details
| Gene Symbol: COL27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032888.3 | NP_116277.2 | 61 | 7120 | ||
| XM_006717308.2 | XP_006717371.1 | 60 | 7134 | |||
| XM_006717310.3 | XP_006717373.1 | 59 | 4933 | |||
| XM_011519138.2 | XP_011517440.1 | 61 | 6752 | |||
| XM_011519140.1 | XP_011517442.1 | 60 | 7134 | |||
| XM_011519145.2 | XP_011517447.1 | 48 | 4362 | |||

COL27A1
Genomic Map