siRNA Details
| Gene Symbol: COL27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032888.3 | NP_116277.2 | 61 | 6427 | ||
| XM_006717308.2 | XP_006717371.1 | 60 | 6441 | |||
| XM_006717310.3 | XP_006717373.1 | 59 | 4240 | |||
| XM_011519138.2 | XP_011517440.1 | 61 | 6059 | |||
| XM_011519140.1 | XP_011517442.1 | 60 | 6441 | |||
| XM_011519145.2 | XP_011517447.1 | 48 | 3669 | |||
| GenBank mRNA | AB058773.1 | - | 58 | 4914 | ||
| AK021957.1 | - | 3 | 1172 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR605514.1 | - | 1 | 137 | Chr. 9 - 114310997 - 114312467 [+] on Build GRCh38 | |

COL27A1
Genomic Map