siRNA Details
| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC017752.1 | - | 3 | 1080 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000458492.1 | - | 3 | 1080 | Chr. 17 - 14210488 - 14217922 [+] on Build GRCh38 | |

COX10
Genomic Map