siRNA Details
| Gene Symbol: SLC4A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 10111 | ||
| NM_001258401.2 | NP_001245330.1 | 25 | 10057 | |||
| XM_011539010.1 | XP_011537312.1 | 26 | 10166 | |||
| XM_011539011.1 | XP_011537313.1 | 26 | 10111 | |||
| XM_011539014.2 | XP_011537316.1 | 16 | 8764 | |||
| XM_017020240.1 | XP_016875729.1 | 26 | 10193 | |||
| XM_017020242.1 | XP_016875731.1 | 26 | 10180 | |||
| GenBank mRNA | AK056222.1 | - | 1 | 954 | ||
| AK124576.1 | - | 1 | 3309 | |||
| AL137310.1 | - | 1 | 2065 | |||

SLC4A8
Genomic Map