siRNA Details
| Gene Symbol: KIAA0509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB007978.1 | - | 1 | 3178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0509| Gene Symbol: KIAA0509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB007978.1 | - | 1 | 3178 | ||