siRNA Details
| Gene Symbol: WDFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534914.1 | XP_011533216.1 | 12 | 2971 | ||
| XM_011534915.2 | XP_011533217.1 | 11 | 5779 | |||
| XM_017020386.1 | XP_016875875.1 | 12 | 11503 | |||
| GenBank mRNA | AK095358.1 | - | 1 | 86 | ||

WDFY2
Genomic Map