siRNA Details
| Gene Symbol: RUSC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093295.1 | - | 3 | 577 | ||
| AK125510.1 | - | 1 | 1949 | |||
| BC131614.1 | - | 2 | 407 | 40115930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RUSC1-AS1| Gene Symbol: RUSC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093295.1 | - | 3 | 577 | ||
| AK125510.1 | - | 1 | 1949 | |||
| BC131614.1 | - | 2 | 407 | 40115930 | ||