siRNA Details
| Gene Symbol: ATP2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833453.1 | - | 1 | 1763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP2C1| Gene Symbol: ATP2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833453.1 | - | 1 | 1763 | ||