siRNA Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042076.1 | - | 2 | 800 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000546686.1 | - | 2 | 800 | Chr. 12 - 52245048 - 52247448 [-] on Build GRCh38 | |

KRT7
Genomic Map