siRNA Details
| Gene Symbol: LINC01285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110393.1 | 1, 2 | 228 | |||
| GenBank mRNA | AK057519.1 | - | 1, 2 | 228 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB026119.1 | - | 1, 2 | 228 | Chr. X - 118839556 - 118855519 [+] on Build GRCh38 | |

LINC01285
Genomic Map