siRNA Details
| Gene Symbol: LOC101929748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040975.2 | - | 1 | 1766 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929748| Gene Symbol: LOC101929748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040975.2 | - | 1 | 1766 | ||