siRNA Details
| Gene Symbol: LOC101928103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110292.1 | 1, 2 | 199 | |||
| GenBank mRNA | AK056813.1 | - | 1, 2 | 199 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA865307.1 | - | 1, 2 | 200 | Chr. 2 - 214810229 - 214962132 [+] on Build GRCh38 | |

LOC101928103
Genomic Map