siRNA Details
| Gene Symbol: LOC101928103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110292.1 | 4 | 1199 | |||
| GenBank mRNA | AK056813.1 | - | 4 | 1199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928103| Gene Symbol: LOC101928103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110292.1 | 4 | 1199 | |||
| GenBank mRNA | AK056813.1 | - | 4 | 1199 | ||