siRNA Details
| Gene Symbol: EBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036592.1 | 2 | 3583 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EBLN3| Gene Symbol: EBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036592.1 | 2 | 3583 | |||