siRNA Details
| Gene Symbol: HOXB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M92299.1 | - | 1 | 109 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DN996030.1 | - | 1 | 142 | Chr. 17 - 48593447 - 48593985 [-] on Build GRCh38 | |

HOXB5
Genomic Map