siRNA Details
| Gene Symbol: SCML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011535703.2 | XP_011534005.1 | 8 | 8658 | ||
| XM_011535704.2 | XP_011534006.1 | 8 | 8536 | |||
| XM_011535706.1 | XP_011534008.1 | 7 | 8502 | |||
| XM_011535709.2 | XP_011534011.1 | 4 | 8692 | |||
| XM_011535710.2 | XP_011534012.1 | 6 | 8369 | |||
| XM_017010677.1 | XP_016866166.1 | 8 | 8827 | |||
| XM_017010678.1 | XP_016866167.1 | 8 | 8740 | |||
| XM_017010679.1 | XP_016866168.1 | 8 | 8638 | |||
| XM_017010680.1 | XP_016866169.1 | 8 | 8706 | |||
| XM_017010681.1 | XP_016866170.1 | 8 | 8616 | |||
| XM_017010682.1 | XP_016866171.1 | 8 | 8841 | |||
| XM_017010683.1 | XP_016866172.1 | 7 | 8499 | |||
| XM_017010685.1 | XP_016866174.1 | 7 | 8626 | |||
| XM_017010686.1 | XP_016866175.1 | 7 | 8611 | |||
| XM_017010687.1 | XP_016866176.1 | 7 | 8537 | |||
| GenBank mRNA | AK128420.1 | - | 1 | 2285 | ||

SCML4
Genomic Map